siRNA Details
| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191772.1 | NP_001178701.1 | 8 | 861 | ||
| XM_006240469.2 | XP_006240531.1 | 7 | 1461 | |||
| XM_006240470.2 | XP_006240532.1 | 7 | 1458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trip11| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191772.1 | NP_001178701.1 | 8 | 861 | ||
| XM_006240469.2 | XP_006240531.1 | 7 | 1461 | |||
| XM_006240470.2 | XP_006240532.1 | 7 | 1458 | |||