siRNA Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240473.2 | XP_006240535.1 | 1 | 428 | ||
| XM_006240474.2 | XP_006240536.1 | 2 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rin3| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240473.2 | XP_006240535.1 | 1 | 428 | ||
| XM_006240474.2 | XP_006240536.1 | 2 | 216 | |||