siRNA Details
| Gene Symbol: Unc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271253.1 | NP_001258182.1 | 37 | 6201 | ||
| XM_006240501.2 | XP_006240563.1 | 38 | 6490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc79| Gene Symbol: Unc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271253.1 | NP_001258182.1 | 37 | 6201 | ||
| XM_006240501.2 | XP_006240563.1 | 38 | 6490 | |||