siRNA Details
| Gene Symbol: Wars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013170.2 | NP_001013188.2 | 7, 8 | 958 | ||
| XM_006240542.2 | XP_006240604.1 | 7, 8 | 995 | |||
| XM_008764811.1 | XP_008763033.1 | 7, 8 | 1149 | |||
| XM_008764812.1 | XP_008763034.1 | 7, 8 | 1746 | |||
| GenBank mRNA | AY724504.1 | - | 7, 8 | 958 | ||
| BC061752.1 | - | 8, 9 | 1021 | 5600013 | ||

Wars
Genomic Map