siRNA Details
| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108061.1 | NP_001101531.1 | 12 | 1384 | ||
| XM_006240588.2 | XP_006240650.1 | 12 | 1364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amn| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108061.1 | NP_001101531.1 | 12 | 1384 | ||
| XM_006240588.2 | XP_006240650.1 | 12 | 1364 | |||