siRNA Details
| Gene Symbol: Ppp1r13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108062.1 | NP_001101532.1 | 10 | 1666 | ||
| XM_006240634.2 | XP_006240696.1 | 12 | 3141 | |||
| XM_008764939.1 | XP_008763161.1 | 12 | 2142 | |||
| XM_008764940.1 | XP_008763162.1 | 11 | 3036 | |||
| XM_008764941.1 | XP_008763163.1 | 11 | 1853 | |||

Ppp1r13b
Genomic Map