siRNA Details
Gene Symbol: Rd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134560.1 | NP_001128032.1 | 3 | 898 | ||
GenBank mRNA | BC158708.1 | - | 3 | 883 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134560.1 | NP_001128032.1 | 3 | 898 | ||
GenBank mRNA | BC158708.1 | - | 3 | 883 |