siRNA Details
Gene Symbol: Gpr132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170595.1 | NP_001164066.1 | 3 | 725 | ||
XM_006240661.2 | XP_006240723.1 | 3 | 1509 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
