siRNA Details
| Gene Symbol: Tmem121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001079243.5 | XP_001079243.2 | 2 | 1079 | ||
| XM_006240674.2 | XP_006240736.1 | 2 | 1055 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem121| Gene Symbol: Tmem121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001079243.5 | XP_001079243.2 | 2 | 1079 | ||
| XM_006240674.2 | XP_006240736.1 | 2 | 1055 | |||