siRNA Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191773.1 | NP_001178702.1 | 16, 17 | 2324 | ||
| XM_006240688.2 | XP_006240750.1 | 18, 19 | 2590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr60| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191773.1 | NP_001178702.1 | 16, 17 | 2324 | ||
| XM_006240688.2 | XP_006240750.1 | 18, 19 | 2590 | |||