siRNA Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191773.1 | NP_001178702.1 | 18 | 2462 | ||
| XM_006240688.2 | XP_006240750.1 | 20 | 2728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr60| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191773.1 | NP_001178702.1 | 18 | 2462 | ||
| XM_006240688.2 | XP_006240750.1 | 20 | 2728 | |||