siRNA Details
| Gene Symbol: Wdr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047889.1 | NP_001041354.1 | 1 | 64 | ||
| XM_006244892.2 | XP_006244954.1 | 3, 4 | 337 | |||
| GenBank mRNA | AY539869.1 | - | 1 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr75| Gene Symbol: Wdr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047889.1 | NP_001041354.1 | 1 | 64 | ||
| XM_006244892.2 | XP_006244954.1 | 3, 4 | 337 | |||
| GenBank mRNA | AY539869.1 | - | 1 | 64 | ||