siRNA Details
| Gene Symbol: Wdr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047889.1 | NP_001041354.1 | 13 | 1573 | ||
| XM_006244892.2 | XP_006244954.1 | 19 | 2161 | |||
| GenBank mRNA | AY539869.1 | - | 13 | 1573 | ||
| FQ234261.1 | - | 6 | 554 | |||

Wdr75
Genomic Map