siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012113.1 | NP_001012113.1 | 11, 12 | 1357 | ||
| GenBank mRNA | BC087129.1 | - | 11, 12 | 1357 | 7112273 | |
| FQ225243.1 | - | 4, 5 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptdss1| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012113.1 | NP_001012113.1 | 11, 12 | 1357 | ||
| GenBank mRNA | BC087129.1 | - | 11, 12 | 1357 | 7112273 | |
| FQ225243.1 | - | 4, 5 | 299 | |||