siRNA Details
| Gene Symbol: Zfp563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134561.3 | NP_001128033.2 | 4 | 321 | ||
| GenBank mRNA | FQ233442.1 | - | 4 | 388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp563| Gene Symbol: Zfp563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134561.3 | NP_001128033.2 | 4 | 321 | ||
| GenBank mRNA | FQ233442.1 | - | 4 | 388 | ||