siRNA Details
Gene Symbol: Zfp563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134561.3 | NP_001128033.2 | 4 | 503 | ||
GenBank mRNA | FQ233442.1 | - | 4 | 570 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134561.3 | NP_001128033.2 | 4 | 503 | ||
GenBank mRNA | FQ233442.1 | - | 4 | 570 |