siRNA Details
| Gene Symbol: Zfp955a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126281.1 | NP_001119753.1 | 4 | 352 | ||
| GenBank mRNA | BC160923.1 | - | 4 | 352 | 8362600 | |
| BC168190.1 | - | 4 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp955a| Gene Symbol: Zfp955a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126281.1 | NP_001119753.1 | 4 | 352 | ||
| GenBank mRNA | BC160923.1 | - | 4 | 352 | 8362600 | |
| BC168190.1 | - | 4 | 320 | |||