siRNA Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033702.1 | NP_001028874.1 | 4 | 381 | ||
| GenBank mRNA | BC099810.1 | - | 4 | 381 | 5621533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf126| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033702.1 | NP_001028874.1 | 4 | 381 | ||
| GenBank mRNA | BC099810.1 | - | 4 | 381 | 5621533 | |