siRNA Details
| Gene Symbol: Wdr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039027.1 | NP_001034116.1 | 2 | 306 | ||
| XM_006240906.1 | XP_006240968.1 | 2 | 383 | |||
| GenBank mRNA | BC099828.1 | - | 2 | 306 | 7934398 | |
| FQ212635.1 | - | 2 | 309 | |||
| FQ212816.1 | - | 2 | 309 | |||
| FQ219525.1 | - | 2 | 309 | |||

Wdr18
Genomic Map