siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108068.1 | NP_001101538.1 | 11, 12 | 1348 | ||
| XM_006240908.2 | XP_006240970.1 | 10, 11 | 2630 | |||
| XM_006240909.1 | XP_006240971.1 | 8, 9 | 2091 | |||
| XM_008765094.1 | XP_008763316.1 | 10, 11 | 1213 | |||
| XM_008765095.1 | XP_008763317.1 | 7, 8 | 2776 | |||

Sbno2
Genomic Map