siRNA Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191577.1 | NP_001178506.1 | 3 | 370 | ||
| XM_006240911.2 | XP_006240973.1 | 4 | 2027 | |||
| XM_006240912.2 | XP_006240974.1 | 4 | 1754 | |||
| XM_006240913.2 | XP_006240975.1 | 4 | 2027 | |||
| XM_006240914.2 | XP_006240976.1 | 4 | 2027 | |||
| XM_008765097.1 | XP_008763319.1 | 3 | 944 | |||

Midn
Genomic Map