siRNA Details
| Gene Symbol: Plekhj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108072.2 | NP_001101542.1 | 6 | 720 | ||
| XM_006240928.1 | XP_006240990.1 | 5 | 814 | |||
| XM_006240929.1 | XP_006240991.1 | 5 | 778 | |||
| XM_006240930.1 | XP_006240992.1 | 6 | 711 | |||
| GenBank mRNA | BC088451.1 | - | 2 | 1908 | ||
| BC168179.1 | - | 6 | 720 | 9092463 | ||
| FQ230285.1 | - | 6 | 989 | |||

Plekhj1
Genomic Map