siRNA Details
| Gene Symbol: Pip5k1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009967.2 | NP_001009967.2 | 7 | 770 | ||
| NM_001033970.1 | NP_001029142.1 | 7 | 770 | |||
| XM_006240935.2 | XP_006240997.1 | 7 | 813 | |||
| XM_008765101.1 | XP_008763323.1 | 7 | 813 | |||
| XM_008765102.1 | XP_008763324.1 | 7 | 813 | |||
| XM_008765103.1 | XP_008763325.1 | 7 | 813 | |||
| GenBank mRNA | AY850259.1 | - | 7 | 709 | ||
| BC101909.1 | - | 7 | 742 | 7933923 | ||

Pip5k1c
Genomic Map