siRNA Details
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240941.2 | XP_006241003.2 | 11 | 1159 | ||
XM_006240942.2 | XP_006241004.2 | 11 | 1132 | |||
XM_008765104.1 | XP_008763326.1 | 10 | 968 | |||
XM_008765105.1 | XP_008763327.1 | 11 | 1157 |
