siRNA Details
| Gene Symbol: RGD1309995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241205.2 | XP_006241267.1 | 16 | 1575 | ||
| XM_008765293.1 | XP_008763515.1 | 9 | 980 | |||
| XM_235003.8 | XP_235003.3 | 16 | 1575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309995| Gene Symbol: RGD1309995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241205.2 | XP_006241267.1 | 16 | 1575 | ||
| XM_008765293.1 | XP_008763515.1 | 9 | 980 | |||
| XM_235003.8 | XP_235003.3 | 16 | 1575 | |||