siRNA Details
| Gene Symbol: RGD1309995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241205.2 | XP_006241267.1 | 27 | 2861 | ||
| XM_008765293.1 | XP_008763515.1 | 20 | 2266 | |||
| XM_235003.8 | XP_235003.3 | 27 | 2861 | |||
| GenBank mRNA | FQ222739.1 | - | 3 | 218 | ||

RGD1309995
Genomic Map