siRNA Details
| Gene Symbol: Actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108081.1 | NP_001101551.1 | 4 | 353 | ||
| XM_006241243.1 | XP_006241305.1 | 4 | 362 | |||
| GenBank mRNA | BC166860.1 | - | 4 | 329 | 9101114 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actr6| Gene Symbol: Actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108081.1 | NP_001101551.1 | 4 | 353 | ||
| XM_006241243.1 | XP_006241305.1 | 4 | 362 | |||
| GenBank mRNA | BC166860.1 | - | 4 | 329 | 9101114 | |