siRNA Details
| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271371.1 | NP_001258300.1 | 13 | 2192 | ||
| XM_008765239.1 | XP_008763461.1 | 1 | 573 | |||
| XM_008765241.1 | XP_008763463.1 | 1 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anks1b| Gene Symbol: Anks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271371.1 | NP_001258300.1 | 13 | 2192 | ||
| XM_008765239.1 | XP_008763461.1 | 1 | 573 | |||
| XM_008765241.1 | XP_008763463.1 | 1 | 574 | |||