siRNA Details
| Gene Symbol: Tmcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108084.1 | NP_001101554.1 | 2 | 603 | ||
| XM_006241255.2 | XP_006241317.1 | 2 | 583 | |||
| XM_006241261.2 | XP_006241323.1 | 3 | 778 | |||
| XM_006241263.2 | XP_006241325.1 | 2 | 1426 | |||

Tmcc3
Genomic Map