siRNA Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003695.2 | NP_003686.1 | 3 | 747 | ||
| GenBank mRNA | BC031330.1 | - | 3 | 747 | 5428818 | |
| BC034542.1 | - | 1 | 1368 | |||
| X82693.1 | - | 3 | 683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY6D| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003695.2 | NP_003686.1 | 3 | 747 | ||
| GenBank mRNA | BC031330.1 | - | 3 | 747 | 5428818 | |
| BC034542.1 | - | 1 | 1368 | |||
| X82693.1 | - | 3 | 683 | |||