siRNA Details
| Gene Symbol: LOC100911483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277222.1 | NP_001264151.1 | 4 | 375 | ||
| GenBank mRNA | FM065403.1 | - | 4 | 371 | ||
| FQ211515.1 | - | 4 | 443 | |||
| FQ221417.1 | - | 4 | 380 | |||
| FQ223566.1 | - | 4 | 386 | |||
| FQ223617.1 | - | 4 | 385 | |||
| FQ223934.1 | - | 4 | 400 | |||
| FQ224478.1 | - | 4 | 388 | |||

LOC100911483
Genomic Map