siRNA Details
| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135858.1 | NP_001129330.1 | 6 | 871 | ||
| XM_008765316.1 | XP_008763538.1 | 5 | 781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmtc3| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135858.1 | NP_001129330.1 | 6 | 871 | ||
| XM_008765316.1 | XP_008763538.1 | 5 | 781 | |||