siRNA Details
| Gene Symbol: OR6A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003696.2 | NP_003687.2 | 1 | 284 | ||
| GenBank mRNA | BC101489.1 | - | 1 | 159 | 8068995 | |
| BC101491.1 | - | 1 | 159 | 8068997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6A2| Gene Symbol: OR6A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003696.2 | NP_003687.2 | 1 | 284 | ||
| GenBank mRNA | BC101489.1 | - | 1 | 159 | 8068995 | |
| BC101491.1 | - | 1 | 159 | 8068997 | ||