siRNA Details
Gene Symbol: Acss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241302.2 | XP_006241364.1 | 12, 13 | 1668 | ||
XM_006241303.2 | XP_006241365.1 | 12, 13 | 2357 | |||
XM_008765327.1 | XP_008763549.1 | 12, 13 | 1408 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
