siRNA Details
| Gene Symbol: Nav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191782.1 | NP_001178711.1 | 34 | 6616 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nav3| Gene Symbol: Nav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191782.1 | NP_001178711.1 | 34 | 6616 | ||