siRNA Details
Gene Symbol: LOC100911101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750322.3 | XP_003750370.1 | 1 | 503 |
Gene Symbol: Lrrc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108096.1 | NP_001101566.1 | 1 | 488 |
