siRNA Details
| Gene Symbol: Cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108098.1 | NP_001101568.1 | 8 | 990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpm| Gene Symbol: Cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108098.1 | NP_001101568.1 | 8 | 990 | ||