siRNA Details
| Gene Symbol: Mon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040176.2 | NP_001035266.2 | 13 | 1752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mon2| Gene Symbol: Mon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040176.2 | NP_001035266.2 | 13 | 1752 | ||