siRNA Details
| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025701.1 | NP_001020872.1 | 6 | 828 | ||
| GenBank mRNA | BC099200.1 | - | 6 | 828 | 7384021 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad21| Gene Symbol: Rad21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025701.1 | NP_001020872.1 | 6 | 828 | ||
| GenBank mRNA | BC099200.1 | - | 6 | 828 | 7384021 | |