siRNA Details
Gene Symbol: RGD1310852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025007.1 | NP_001020178.1 | 6 | 1102 | ||
GenBank mRNA | BC091232.1 | - | 6 | 1102 | 7322456 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029418, AK209094, AK089715, AK018655, BC050895, and others.
Panther Classification:
