siRNA Details
Gene Symbol: Fbxo43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012117.1 | NP_001012117.2 | 5 | 2106 | ||
XM_006241535.2 | XP_006241597.1 | 5 | 1864 | |||
GenBank mRNA | BC082107.1 | - | 5 | 2106 | 7134870 |
