siRNA Details
| Gene Symbol: Osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012118.1 | NP_001012118.1 | 2 | 507 | ||
| XM_006241542.2 | XP_006241604.1 | 2 | 548 | |||
| GenBank mRNA | BC079211.1 | - | 2 | 507 | 7131819 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osr2| Gene Symbol: Osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012118.1 | NP_001012118.1 | 2 | 507 | ||
| XM_006241542.2 | XP_006241604.1 | 2 | 548 | |||
| GenBank mRNA | BC079211.1 | - | 2 | 507 | 7131819 | |