siRNA Details
| Gene Symbol: Pop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241543.2 | XP_006241605.1 | 11, 12 | 1733 | ||
| GenBank mRNA | FQ223904.1 | - | 11, 12 | 1708 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pop1| Gene Symbol: Pop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241543.2 | XP_006241605.1 | 11, 12 | 1733 | ||
| GenBank mRNA | FQ223904.1 | - | 11, 12 | 1708 | ||