siRNA Details
| Gene Symbol: Laptm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013174.1 | NP_001013192.1 | 4 | 387 | ||
| GenBank mRNA | BC086442.1 | - | 4 | 387 | 7303600 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Laptm4b| Gene Symbol: Laptm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013174.1 | NP_001013192.1 | 4 | 387 | ||
| GenBank mRNA | BC086442.1 | - | 4 | 387 | 7303600 | |