siRNA Details
| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134880.1 | NP_001128352.1 | 1, 2 | 166 | ||
| GenBank mRNA | FQ209946.1 | - | 1, 2 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrac1| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134880.1 | NP_001128352.1 | 1, 2 | 166 | ||
| GenBank mRNA | FQ209946.1 | - | 1, 2 | 167 | ||