siRNA Details
Gene Symbol: Naprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207609.1 | NP_997492.1 | 9 | 1230 | ||
GenBank mRNA | AY214330.1 | - | 9 | 1230 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Naprt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207609.1 | NP_997492.1 | 9 | 1230 | ||
GenBank mRNA | AY214330.1 | - | 9 | 1230 |