siRNA Details
Gene Symbol: RGD1311188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135897.1 | NP_001129369.1 | 2 | 345 | ||
XM_008765570.1 | XP_008763792.1 | 2 | 316 | |||
GenBank mRNA | BC162017.1 | - | 2 | 345 | 9098233 |
Gene Symbol: Zfp707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135903.1 | NP_001129375.1 | 2 | 339 | ||
XM_006241815.2 | XP_006241877.1 | 2 | 569 | |||
XM_006241816.2 | XP_006241878.1 | 2 | 567 | |||
GenBank mRNA | BC161959.1 | - | 2 | 339 | 9027463 |
