siRNA Details
| Gene Symbol: Maf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014085.1 | NP_001014107.1 | 5, 6 | 863 | ||
| XM_006241811.1 | XP_006241873.1 | 5, 6 | 938 | |||
| XM_006241812.1 | XP_006241874.1 | 5, 6 | 907 | |||
| GenBank mRNA | BC083712.1 | - | 5, 6 | 863 | 7190634 | |

Maf1
Genomic Map