siRNA Details
| Gene Symbol: RGD1561328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025122.2 | NP_001020293.2 | 2 | 1165 | ||
| GenBank mRNA | BC091178.1 | - | 2 | 1165 | 7325408 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561328| Gene Symbol: RGD1561328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025122.2 | NP_001020293.2 | 2 | 1165 | ||
| GenBank mRNA | BC091178.1 | - | 2 | 1165 | 7325408 | |