siRNA Details
| Gene Symbol: Apol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241942.2 | XP_006242004.1 | 3, 4 | 831 | ||
| XM_006241943.2 | XP_006242005.1 | 4, 5 | 713 | |||
| XM_006241944.2 | XP_006242006.1 | 3, 4 | 548 | |||
| XM_006241945.1 | XP_006242007.1 | 3, 4 | 284 | |||
| XM_006241946.2 | XP_006242008.1 | 3, 4 | 528 | |||
| XM_006241947.2 | XP_006242009.1 | 4, 5 | 604 | |||
| XM_006241948.2 | XP_006242010.1 | 4, 5 | 576 | |||
| XM_008765644.1 | XP_008763866.1 | 3, 4 | 304 | |||
| XM_008765645.1 | XP_008763867.1 | 4, 5 | 556 | |||
| XM_235465.8 | XP_235465.6 | 4, 5 | 734 | |||
| Gene Symbol: LOC100911804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750381.3 | XP_003750429.1 | 735 | |||
| XM_006241953.2 | XP_006242015.1 | 832 | ||||
| XM_006241954.2 | XP_006242016.1 | 714 | ||||
| XM_006241955.2 | XP_006242017.1 | 3, 4 | 549 | |||
| XM_006241956.1 | XP_006242018.1 | 284 | ||||
| XM_006241957.2 | XP_006242019.1 | 3, 4 | 529 | |||
| XM_006241958.2 | XP_006242020.1 | 4, 5 | 605 | |||
| XM_006241959.2 | XP_006242021.1 | 4, 5 | 577 | |||
| XM_008765799.1 | XP_008764021.1 | 304 | ||||
| XM_008765800.1 | XP_008764022.1 | 4, 5 | 557 | |||

Apol2
Genomic Map